5-Norbornen-2-yl acetate, mixture of endo and exo, 98%
5-Norbornen-2-yl acetate, mixture of endo and exo, 98%
5-Norbornen-2-yl acetate, mixture of endo and exo, 98%
Thermo Scientific Chemicals

5-Norbornen-2-yl acetate, mixture of endo and exo, 98%

CAS: 6143-29-9 | C9H12O2 | 152.193 g/mol
Quantity:
25 g
100 g
Catalog number H56907.22
also known as H56907-22
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS6143-29-9
IUPAC Namebicyclo[2.2.1]hept-5-en-2-yl acetate
Molecular FormulaC9H12O2
InChI KeyDRWRVXAXXGJZIO-UHFFFAOYNA-N
SMILESCC(=O)OC1CC2CC1C=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only