2,3-Dimethylbenzaldehyde, 97%
2,3-Dimethylbenzaldehyde, 97%
2,3-Dimethylbenzaldehyde, 97%
2,3-Dimethylbenzaldehyde, 97%
Thermo Scientific Chemicals

2,3-Dimethylbenzaldehyde, 97%

CAS: 5779-93-1 | C9H10O | 134.178 g/mol
Quantity:
5 g
25 g
100 g
Catalog number H64425.14
also known as H64425-14
Price (JPY)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS5779-93-1
IUPAC Name2,3-dimethylbenzaldehyde
Molecular FormulaC9H10O
InChI KeyUIFVCPMLQXKEEU-UHFFFAOYSA-N
SMILESCC1=CC=CC(C=O)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only