Methyl 5-chloropyrazine-2-carboxylate, 98%, Thermo Scientific Chemicals
Methyl 5-chloropyrazine-2-carboxylate, 98%, Thermo Scientific Chemicals
Methyl 5-chloropyrazine-2-carboxylate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 5-chloropyrazine-2-carboxylate, 98%, Thermo Scientific Chemicals

Quantity:
1 g
5 g
Catalog number H66306.03
also known as H66306-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Beilstein880046
CAS33332-25-1
Chemical Name or MaterialMethyl 5-chloropyrazine-2-carboxylate
Formula Weight172.57
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only