Fluorolink∣r E10-H
Thermo Scientific Chemicals

Fluorolink∣r E10-H

Use in polymer modifications | CAS: 162492-15-1
Quantity:
25 g
100 g
Catalog number H66374.22
also known as H66374-22
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS162492-15-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear
FormLiquid
Water Content≤500 ppm
Color (APHA)≤100
Equivalent Weight800-1100 AMW
Use in polymer modifications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Use in polymer modifications.

Solubility
Soluble in fluorinated solvents, alcohol, and ketones. Insoluble in water.

Notes
Incompatible with heat and oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only
Federico Persico; Maurizio Sansotera; Maria Vittoria Diamanti; Luca Magagnin; Francesco Venturini; Walter Navarrini. Effect of amorphous fluorinated coatings on photocatalytic properties of anodized titanium surfaces. Thin Solid Films.2013, 545, 210-216.

Frequently asked questions (FAQs)

What applications does Fluorolink E10-H have?

Thanks to its functional hydroxyl-end groups, Fluorolink E10-H can be used to modify polymers such as urethanes, esters, epoxies and acrylates. Fluorolink E10-H is especially suitable as a building block and polymer modifier for those applications that require a combination of excellent water and oil repellency, easy removal of graffiti and fingerprints, low friction, low reflectivity (low refractive index) and high chemical resistance.

What is the chemical structure of Fluorolink E10-H?

Fluorolink E10-H is a dialcohol-terminated, ethoxylated perfluoropolyether (PFPE) having the following chemical structure: HO(CH2CH2O)nCH2CF2O(CF2CF2O)p(CF2O)qCF2CH.

What is the decomposition temperature of Fluorolink E10-H?

Fluorolink E10-H decomposes at temperatures > 250°C.