N-Fmoc-O-trityl-D-serine, 95%
N-Fmoc-O-trityl-D-serine, 95%
N-Fmoc-O-trityl-D-serine, 95%
Thermo Scientific Chemicals

N-Fmoc-O-trityl-D-serine, 95%

CAS: 212688-51-2 | C37H31NO5 | 569.657 g/mol
Quantity:
5 g
25 g
Catalog number H66515.06
also known as H66515-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Specifications
CAS212688-51-2
Chemical Name or MaterialN-Fmoc-O-trityl-D-serine
Formula Weight569.66
IUPAC Name(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-trityloxypropanoic acid
InChI KeyUCARTONYOJORBQ-UUWRZZSWSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only