1-(2,3-Epoxypropyl)-2-nitroimidazole, 97%
1-(2,3-Epoxypropyl)-2-nitroimidazole, 97%
1-(2,3-Epoxypropyl)-2-nitroimidazole, 97%
Thermo Scientific Chemicals

1-(2,3-Epoxypropyl)-2-nitroimidazole, 97%

CAS: 13551-90-1 | C6H7N3O3 | 169.14 g/mol
Quantity:
250 mg
1 g
Catalog number H66536.MD
also known as H66536-MD
Price (JPY)
-
Quantity:
250 mg
Request bulk or custom format
Specifications
Beilstein612095
CAS13551-90-1
Chemical Name or Material1-(2,3-Epoxypropyl)-2-nitroimidazole
Formula Weight169.14
Health Hazard 1H302-H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only