6-Chloroindole-2-carboxylic acid, 96%, Thermo Scientific Chemicals
6-Chloroindole-2-carboxylic acid, 96%, Thermo Scientific Chemicals
6-Chloroindole-2-carboxylic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Chloroindole-2-carboxylic acid, 96%, Thermo Scientific Chemicals

Quantity:
1 g
5 g
Catalog number H66864.06
also known as H66864-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS16732-75-5
IUPAC Name6-chloro-1H-indole-2-carboxylic acid
Molecular FormulaC9H6ClNO2
InChI KeyBKPSJOSKWKTWAG-UHFFFAOYSA-N
SMILESOC(=O)C1=CC2=CC=C(Cl)C=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMRConforms
FormPowder
Assay (HPLC)≥95.0%
Appearance (Color)White to pale cream.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only