4-Aminoquinazoline, 97%, Thermo Scientific Chemicals
4-Aminoquinazoline, 97%, Thermo Scientific Chemicals
4-Aminoquinazoline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Aminoquinazoline, 97%, Thermo Scientific Chemicals

Quantity:
1 g
5 g
Catalog number H66948.03
also known as H66948-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Beilstein118621
CAS15018-66-3
Chemical Name or Material4-Aminoquinazoline
Formula Weight145.06
Health Hazard 1H302-H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only