Buffer Solution pH 2.00 (Glycine), NIST Standard Solution Ready to Use for pH Measurement, Fisher Chemical™
Fisher Chemical™

Buffer Solution pH 2.00 (Glycine), NIST Standard Solution Ready to Use for pH Measurement, Fisher Chemical™

Catalog NumberQuantity
J-2800-15
also known as J/2800/15
1 L
Catalog number J-2800-15
also known as J/2800/15
Price (JPY)
-
Quantity:
1 L
Request bulk or custom format
Specifications
ColorColorless
MDL Number217672
PackagingHDPE plastic bottle
Physical FormLiquid
Quantity1 L
View more
Buffer Solution pH 2.00 (Glycine), NIST Standard Solution Ready to Use for pH Measurement