Buffer Solution pH5.00 (Phthalate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Buffer Solution pH5.00 (Phthalate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Buffer Solution pH5.00 (Phthalate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Fisher Chemical™

Buffer Solution pH5.00 (Phthalate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™

Catalog NumberQuantity
J-2830-08
also known as J/2830/08
500 mL
Catalog number J-2830-08
also known as J/2830/08
Price (JPY)
-
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS877-24-7
Specifications
CAS877-24-7
ColorColorless
MDL Number13070
PackagingHDPE plastic bottle
Physical FormLiquid
View more
Buffer Solution pH5.00 (Phthalate), NIST Standard Solution ready to use for pH measurement