Buffer Colour Coded Solution pH10.00 (Borate) Blue, NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Buffer Colour Coded Solution pH10.00 (Borate) Blue, NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Buffer Colour Coded Solution pH10.00 (Borate) Blue, NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Fisher Chemical™

Buffer Colour Coded Solution pH10.00 (Borate) Blue, NIST Standard Solution ready to use for pH measurement, Fisher Chemical™

Buffer Colour Coded Solution pH10.00 (Borate) Blue, NIST Standard Solution ready to use for pH measurement, Fisher Chemical, Quantity: 1L, Packaging: HDPE plastic bottle, pH: 10, MDL Number: 163147, Physical Form: Liquid
Catalog NumberQuantity
J-2885-15
also known as J/2885/15
1 L
Catalog number J-2885-15
also known as J/2885/15
Price (JPY)
-
Quantity:
1 L
Request bulk or custom format
Specifications
CAS10043-35-3
ColorBlue
FormulationPotassium Chloride, Water, Trisodium Orthoborate, Thymol Blue, Boric Acid (H3BO3)
IUPAC Nameboric acid
InChI KeyKGBXLFKZBHKPEV-UHFFFAOYSA-N
View more
Buffer Colour Coded Solution pH10.00 (Borate) Blue, NIST Standard Solution ready to use for pH measurement, Fisher Chemical, Quantity: 1L, Packaging: HDPE plastic bottle, pH: 10, MDL Number: 163147, Physical Form: Liquid
Ready to use for pH measurement.
Each solution is easily identified by its color, saving time and reducing errors.
Our buffer solutions are a high quality and reliable range of products that ensure accurate results for calibration/titration.
  • Standard and color-coded buffer solutions
  • Manufactured to a tolerance of ±0.02 pH units
  • Accurately standardized and labelled with lot-specific pH at 20°C
  • All buffers are traceable to NIST and BS 1647 standard
  • Packed in HDPE containers to help protect the products integrity an ensure safe handling (refers to 1L, 2.5L, and 5L quantities)
  • Standard buffers are available over the range pH2 to pH10
  • Color-coded buffer solutions are available over the range pH4, pH7, and pH