Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Fisher Chemical™

Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

ClH, CAS Number-7647-01-0, hydrogen chloride, hydrochloric acid, acide chlorhydrique, chlorwasserstoff, anhydrous hydrochloric acid, chlorohydric acid, chloorwaterstof, hydrogen chloride hcl, muriatic acid, spirits of salt, 10L, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol
Catalog NumberQuantityPackaging
J-4350-24
also known as J/4350/24
10 LBag in box
Catalog number J-4350-24
also known as J/4350/24
Price (JPY)
-
Quantity:
10 L
Packaging:
Bag in box
Request bulk or custom format
Specifications
Boiling Point100°C
CAS7647-01-0
ChEBICHEBI:17883
ColorColorless
Formula Weight36.46g/mol
View more
ClH, CAS Number-7647-01-0, hydrogen chloride, hydrochloric acid, acide chlorhydrique, chlorwasserstoff, anhydrous hydrochloric acid, chlorohydric acid, chloorwaterstof, hydrogen chloride hcl, muriatic acid, spirits of salt, 10L, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol