Potassium Dichromate Solution 0.0167M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Potassium Dichromate Solution 0.0167M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Potassium Dichromate Solution 0.0167M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Fisher Chemical™

Potassium Dichromate Solution 0.0167M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™

Cr2K2O7, CAS Number-7778-50-9, dipotassium bichromate, bichromate of potash, dichromic acid dipotassium salt, potassium dichromate, potassium dichromate vi, iopezite, dipotassium dichromium heptaoxide, kaliumdichromat, potassium bichromate, dipotassium dichromate, 1AMP, CHEBI:53444, Orange
Catalog NumberQuantity
J-6212C-05
also known as J/6212C/05
1 AMP
Catalog number J-6212C-05
also known as J/6212C/05
Price (JPY)
-
Quantity:
1 AMP
Request bulk or custom format
Specifications
CAS7778-50-9
ChEBICHEBI:53444
ColorOrange
IUPAC Namedipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
InChI KeyKMUONIBRACKNSN-UHFFFAOYSA-N
View more
Cr2K2O7, CAS Number-7778-50-9, dipotassium bichromate, bichromate of potash, dichromic acid dipotassium salt, potassium dichromate, potassium dichromate vi, iopezite, dipotassium dichromium heptaoxide, kaliumdichromat, potassium bichromate, dipotassium dichromate, 1AMP, CHEBI:53444, Orange