Sodium Hydroxide Solution 2M (2N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Fisher Chemical™
Sodium Hydroxide Solution 2M (2N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Fisher Chemical™
Sodium Hydroxide Solution 2M (2N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Fisher Chemical™
Fisher Chemical™

Sodium Hydroxide Solution 2M (2N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Fisher Chemical™

Catalog NumberQuantityPackaging
J-7610-17
also known as J/7610/17
2.5 LHDPE plastic bottle
Catalog number J-7610-17
also known as J/7610/17
Price (JPY)
-
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Specifications
Boiling Point105°C
CAS1310-73-2
ChEBICHEBI:32145
ColorColorless
IUPAC Namesodium;hydroxide
View more
Sodium Hydroxide Solution 2M (2N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur.