Sodium Hydroxide Solution 0.1M (0.1N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Sodium Hydroxide Solution 0.1M (0.1N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Sodium Hydroxide Solution 0.1M (0.1N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Fisher Chemical™

Sodium Hydroxide Solution 0.1M (0.1N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

HNaO, CAS Number-1310-73-2, aetznatron, ascarite, caustic soda, soda, caustic, soda lye, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic, 1L, 100 deg.C, CHEBI:32145, Colorless, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 3548, 39.997 | CAS: 1310-73-2 | HNaO | 39.997 g/mol
Quantity:
1 L
2.5 L
Packaging:
HDPE plastic bottle
Catalog number J-7660-15
also known as J/7660/15
Price (JPY)
-
Quantity:
1 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Specifications
Boiling Point100°C
CAS1310-73-2
ChEBICHEBI:32145
ColorColorless
IUPAC Namesodium;hydroxide
View more
HNaO, CAS Number-1310-73-2, aetznatron, ascarite, caustic soda, soda, caustic, soda lye, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic, 1L, 100 deg.C, CHEBI:32145, Colorless, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 3548, 39.997