Sodium Hydroxide Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Sodium Hydroxide Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Sodium Hydroxide Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Sodium Hydroxide Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Fisher Chemical™

Sodium Hydroxide Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™

HNaO, CAS Number-1310-73-2, sodium hydroxide, sodium hydroxide na oh, soda lye, caustic soda, ascarite, white caustic, sodium hydrate, sodium hydroxide solution, soda, caustic, aetznatron, 6 ampules, 100 deg.C, CHEBI:32145, Colorless, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 3548, 39.997
Quantity:
6 Ampules
1 Ampule
Catalog number J-7660C-90
also known as J/7660C/90
Price (JPY)
-
Contact Us ›
Quantity:
6 Ampules
Request bulk or custom format
Specifications
Boiling Point100°C
CAS1310-73-2
ChEBICHEBI:32145
ColorColorless
IUPAC Namesodium;hydroxide
View more
HNaO, CAS Number-1310-73-2, sodium hydroxide, sodium hydroxide na oh, soda lye, caustic soda, ascarite, white caustic, sodium hydrate, sodium hydroxide solution, soda, caustic, aetznatron, 6 ampules, 100 deg.C, CHEBI:32145, Colorless, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 3548, 39.997