Sodium Hydroxide Solution 10M (10N) ≈40 (w/v), Standard Solution Ready To Use, for Volumetric Analysis
Sodium Hydroxide Solution 10M (10N) ≈40 (w/v), Standard Solution Ready To Use, for Volumetric Analysis
Sodium Hydroxide Solution 10M (10N) ≈40 (w/v), Standard Solution Ready To Use, for Volumetric Analysis
Fisher Chemical™

Sodium Hydroxide Solution 10M (10N) ≈40 (w/v), Standard Solution Ready To Use, for Volumetric Analysis

Catalog NumberQuantityPackaging
J-7800-15
also known as J/7800/15
1 LHDPE plastic bottle
Catalog number J-7800-15
also known as J/7800/15
Price (JPY)
-
Quantity:
1 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Specifications
Boiling Point120°C
CAS1310-73-2
ChEBICHEBI:32145
IUPAC Namesodium;hydroxide
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
View more
Sodium Hydroxide Solution 10M (10N) ≈40 (w/v), Standard Solution Ready To Use, for Volumetric Analysis