Aluminum Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Aluminum Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Aluminum Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Fisher Chemical™

Aluminum Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™

Quantity:
100 mL
500 mL
Catalog number J-8000-08
also known as J/8000/08
Price (JPY)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
CAS13473-90-0
ColorColorless
MDL Number134029
PackagingHDPE plastic bottle
Physical FormLiquid
View more
Aluminum Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M