Sulfuric Acid Solution 2M (4N), Pure, Bench Reagent, Fisher Chemical™
Sulfuric Acid Solution 2M (4N), Pure, Bench Reagent, Fisher Chemical™
Sulfuric Acid Solution 2M (4N), Pure, Bench Reagent, Fisher Chemical™
Fisher Chemical™

Sulfuric Acid Solution 2M (4N), Pure, Bench Reagent, Fisher Chemical™

H2O4S, CAS Number-7664-93-9, acide sulfurique, battery acid, vitriol brown oil, electrolyte acid, dipping acid, acidum sulfuricum, sulphuric acid, mattling acid, dihydrogen sulfate, oil of vitriol, 1L, 120 deg.C, CHEBI:26836, Colorless, QAOWNCQODCNURD-UHFFFAOYSA-N, sulfuric acid, 64589, 98.072
Catalog NumberQuantityPackaging
J-8410-PB15
also known as J/8410/PB15
1 LHDPE plastic bottle
Catalog number J-8410-PB15
also known as J/8410/PB15
Price (JPY)
-
Quantity:
1 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Specifications
Boiling Point120°C
CAS7664-93-9
ChEBICHEBI:26836
Chemical Name or MaterialSulfuric Acid Solution
ColorColorless
View more
H2O4S, CAS Number-7664-93-9, acide sulfurique, battery acid, vitriol brown oil, electrolyte acid, dipping acid, acidum sulfuricum, sulphuric acid, mattling acid, dihydrogen sulfate, oil of vitriol, 1L, 120 deg.C, CHEBI:26836, Colorless, QAOWNCQODCNURD-UHFFFAOYSA-N, sulfuric acid, 64589, 98.072