7-Chlorokynurenic acid, 98+%
7-Chlorokynurenic acid, 98+%
7-Chlorokynurenic acid, 98+%
Thermo Scientific Chemicals

7-Chlorokynurenic acid, 98+%

NMDA receptor antagonist at the glycine site | CAS: 18000-24-3 | C10H6ClNO3 | 223.61 g/mol
Quantity:
10 mg
50 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61359.MF
also known as J61359-MF
Price (JPY)
-
Quantity:
50 mg
Request bulk or custom format
Specifications
CAS18000-24-3
ChEBICHEBI:63965
Chemical Name or Material7-Chlorokynurenic acid
ColorWhite to Yellow
EINECS Number241-913-6
View more
NMDA receptor antagonist at the glycine site

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only