2'-Deoxyinosine
2'-Deoxyinosine
2'-Deoxyinosine
Thermo Scientific Chemicals

2'-Deoxyinosine

CAS: 890-38-0 | C10H12N4O4 | 252.23 g/mol
Quantity:
10 g
25 g
Catalog number J61411.09
also known as J61411-09
Price (JPY)
-
Request A Quote
Quantity:
10 g
Request bulk or custom format
Specifications
Beilstein33517
CAS890-38-0
ChEBICHEBI:28997
Chemical Name or Material2'-Deoxyinosine
ColorWhite
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only