Cilostazol, 98%, Thermo Scientific Chemicals
Cilostazol, 98%, Thermo Scientific Chemicals
Cilostazol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cilostazol, 98%, Thermo Scientific Chemicals

An inhibitor of phosphodiesterase III
Catalog NumberQuantity
J62301.MA
also known as J62301-MA
10 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62301.MA
also known as J62301-MA
Price (JPY)
-
Quantity:
10 mg
Request bulk or custom format
Specifications
CAS73963-72-1
ChEBICHEBI:31401
Chemical Name or MaterialCilostazol
ColorWhite
Formula Weight369.46
View more
An inhibitor of phosphodiesterase III

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only