Fibrinogen, Bovine
Thermo Scientific Chemicals

Fibrinogen, Bovine

Fibrinogen (CAS # 9001-32-5) is a glycoprotein produced in the vertebrate liver that aids in clot formation.
Quantity:
1g
5g
10g
25g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63276.06
also known as J63276-06
Price (JPY)
-
Quantity:
5g
Request bulk or custom format
Chemical Identifiers
CAS9001-32-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
FormPowder
AssaySolubility: 3% w/v in DIH2O
Assay (unspecified)Solubility: 3% w/v in DIH2O
AssayClottable Protein: ≥90%
View more
A soluble plasma glycoprotein, produced by the liver, that is converted by thrombin into fibrin during the blood coagulation process. Fibrinogen is a natural blood protein involved in the coagulation process. It is used instead of traditional sources of fibrinogen such as the blood products, fresh frozen plasma and cryoprecipitate. It helps in blood clot formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Fibrinogen exists in blood as a dimer of three bonded, non-identical chains, FGA, FGG, and FGB

• Fibrinogen is cleaved by thrombin to form fibrin, the most abundant component of blood clots

Application

• Fibrinogen can be used in a laboratory setting to form polymer hydrogels, particularly for use as a tissue scaffold

• Fibrinogen may be useful in the research of neurodegeneration and brain injury; some evidence shows that it has a signaling role in neural progenitor cells

RUO – Research Use Only

General References:

  1. Jira W, Schwägele F. A sensitive HPLC-MS/MS method for the simultaneous detection of microbial transglutaminase, and bovine and porcine fibrinogen/thrombin in restructured meat. Food Chem. 2017 Dec 15;237:841-848. doi: 10.1016/j.foodchem.2017.06.030. Epub 2017 Jun 12. PMID: 28764076.
  2. Kargl R, Bračič M, Resnik M, Mozetič M, Bauer W, Stana Kleinschek K, Mohan T. Affinity of Serum Albumin and Fibrinogen to Cellulose, Its Hydrophobic Derivatives and Blends. Front Chem. 2019 Sep 6;7:581. doi: 10.3389/fchem.2019.00581. PMID: 31552215; PMCID: PMC6743410.