Calcitonin (8-32), salmon
Calcitonin (8-32), salmon
Calcitonin (8-32), salmon
Thermo Scientific™

Calcitonin (8-32), salmon

Catalog number J64415.LB0
also known as J64415-LB0
Price (JPY)
-
Request bulk or custom format
Chemical Identifiers
CAS155069-90-2
IUPAC Name(4S)-4-{[(1S)-1-{[(1S)-1-{[(1S)-5-amino-1-{[(1S)-1-{[(1S)-1-{[(1S,2R)-1-{[(2S)-1-[(2S)-2-{[(1S)-4-carbamimidamido-1-{[(1S,2R)-1-{[(1S)-2-carbamoyl-1-{[(1S,2R)-1-[({[(1S)-1-[({[(2S,3R)-1-[(2S)-2-carbamoylpyrrolidin-1-yl]-3-hydroxy-1-oxobutan-2-yl]carbamoyl}methyl)carbamoyl]-2-hydroxyethyl]carbamoyl}methyl)carbamoyl]-2-hydroxypropyl]carbamoyl}ethyl]carbamoyl}-2-hydroxypropyl]carbamoyl}butyl]carbamoyl}pyrrolidin-1-yl]-3-(4-hydroxyphenyl)-1-oxopropan-2-yl]carbamoyl}-2-hydroxypropyl]carbamoyl}-3-carbamoylpropyl]carbamoyl}-3-methylbutyl]carbamoyl}pentyl]carbamoyl}-2-(1H-imidazol-4-yl)ethyl]carbamoyl}-3-methylbutyl]carbamoyl}-4-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-6-amino-2-{2-[(2S)-2-[(2S)-2-amino-3-methylbutanamido]-4-methylpentanamido]acetamido}hexanamido]-4-methylpentanamido]-3-hydroxypropanamido]-4-carbamoylbutanamido]butanoic acid
Molecular FormulaC119H198N36O37
InChI KeyCUQBOYYGZCRRKW-LPJWDPCBSA-N
SMILESCC(C)C[C@H](NC(=O)[C@@H](N)C(C)C)C(=O)NCC(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC1=CNC=N1)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CC1=CC=C(O)C=C1)C(=O)N1CCC[C@H]1C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H]([C@@H](C)O)C(=O)NCC(=O)N[C@@H](CO)C(=O)NCC(=O)N[C@@H]([C@@H](C)O)C(=O)N1CCC[C@H]1C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Mass SpecConsistent with expected mass
Appearance (Color)White to pale cream to pale yellow
FormPowder
Assay from Supplier's CofA≥94.0%