Heptyl beta-D-glucopyranoside, 99%
Heptyl beta-D-glucopyranoside, 99%
Heptyl beta-D-glucopyranoside, 99%
Thermo Scientific Chemicals

Heptyl beta-D-glucopyranoside, 99%

CAS: 78617-12-6 | C13H26O6 | 278.345 g/mol
Quantity:
500 mg
1 g
Catalog number J66576.03
also known as J66576-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Beilstein8806
CAS78617-12-6
Chemical Name or MaterialHeptyl beta-D-glucopyranoside
ColorWhite
Formula Weight278.34
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only