Gramicidin, 95%
Thermo Scientific Chemicals

Gramicidin, 95%

For cellular and molecular biology applications | 1882.29 g/mol
Quantity:
100 mg
500 mg
1 g
Catalog number J66771.ME
also known as J66771-ME
Price (JPY)
-
Quantity:
500 mg
Request bulk or custom format
Chemical Identifiers
CAS1405-97-6
SpecificationsSpecification SheetSpecification Sheet
Elemental AnalysisGramicidin A1: ≥60.0%
CommentSource is Bacillus Brevis
Melting Point≥229?C
FormPowder
Elemental AnalysisGramicidin A1, A2, B1, C1 and C2: ≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only
Contents & Storage
2°C to 8°C