Cefsulodin sodium salt, 50 mg/ml in distilled water, sterile-filtered, Thermo Scientific Chemicals
Cefsulodin sodium salt, 50 mg/ml in distilled water, sterile-filtered, Thermo Scientific Chemicals
Cefsulodin sodium salt, 50 mg/ml in distilled water, sterile-filtered, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cefsulodin sodium salt, 50 mg/ml in distilled water, sterile-filtered, Thermo Scientific Chemicals

Quantity:
1 mL
5 x 1 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J67192.8EQ
also known as J67192-8=
Price (JPY)
-
Quantity:
5 x 1 mL
Request bulk or custom format
Specifications
CAS52152-93-9
Chemical Name or MaterialCefsulodin sodium salt
EINECS Number257-692-4
Health Hazard 1H317-H334-H335
Health Hazard 2GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
View more
Cefsulodin is most commonly used in CIN agar to select for Yersinia microorganisms. The compound displays a mechanism of action like many β lactam antibiotics through inhibition of cell wall synthesis by competitively inhibiting penicillin binding protein (PBP) crosslinking of peptidoglycan resulting in inhibition of the final transpeptidation step. Through the inability for Cefsulodin sodium salt hydrate to inhibit cefsulodin-resistant mutants of Pseudomonas aeruginosa PAO4089 growth displayed that Cefsulodin sodium salt hydrate may compete with PBP3 in addition to PBP1A and PBP1B.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cefsulodin is most commonly used in CIN agar to select for Yersinia microorganisms. The compound displays a mechanism of action like many β lactam antibiotics through inhibition of cell wall synthesis by competitively inhibiting penicillin binding protein (PBP) crosslinking of peptidoglycan resulting in inhibition of the final transpeptidation step. Through the inability for Cefsulodin sodium salt hydrate to inhibit cefsulodin-resistant mutants of Pseudomonas aeruginosa PAO4089 growth displayed that Cefsulodin sodium salt hydrate may compete with PBP3 in addition to PBP1A and PBP1B.

Solubility
Soluble in water.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only