A-803467, 98%, Thermo Scientific Chemicals
A-803467, 98%, Thermo Scientific Chemicals
A-803467, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

A-803467, 98%, Thermo Scientific Chemicals

Quantity:
5 mg
50 mg
Catalog number J67281.MF
also known as J67281-MF
Price (JPY)
-
Quantity:
50 mg
Request bulk or custom format
Specifications
CAS944261-79-4
Chemical Name or MaterialA-803467
ColorWhite
Formula Weight357.79
Health Hazard 1H302
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only