Sodium phosphate, 0.2M buffer soln., pH 7.4, low endotoxin
Sodium phosphate, 0.2M buffer soln., pH 7.4, low endotoxin
Sodium phosphate, 0.2M buffer soln., pH 7.4, low endotoxin
Thermo Scientific Chemicals

Sodium phosphate, 0.2M buffer soln., pH 7.4, low endotoxin

CAS: 7732-18-5
Quantity:
100 mL
250 mL
Catalog number J67424.AE
also known as J67424-AE
Price (JPY)
-
Quantity:
100 mL
Request bulk or custom format
Specifications
CAS7732-18-5
ColorColorless
Recommended StorageAmbient Temperatures
Concentration0.2M buffer soln.
Physical FormLiquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only