Mirin, 95%
Mirin, 95%
Mirin, 95%
Thermo Scientific Chemicals

Mirin, 95%

CAS: 299953-00-7 | C10H8N2O2S, C10H8N2O2S | 220.25 g/mol
Quantity:
5 mg
10 mg
50 mg
Catalog number J67462.MA
also known as J67462-MA
Price (JPY)
-
Quantity:
10 mg
Request bulk or custom format
Specifications
Assay95%
CAS299953-00-7
DescriptionOrange
FormSolid
Formula Weight220.25g/mol
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C