Krebs-Heneseleit buffer solution
Thermo Scientific Chemicals
Krebs-Heneseleit buffer solution
Quantity:
500 mL
1 L
Catalog number J67820.AP
also known as J67820-AP
Price (JPY)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
PackagingPlastic Bottle
Recommended StorageAmbient Temperatures
Physical FormLiquid
Quantity500 mL
Unit SizeEach
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Frequently asked questions (FAQs)
What are the concentrations of the components of Krebs-Heneseleit buffer solution?
The Krebs-Heneseleit buffer solution contains:
  • 118 mM sodium chloride
  • 4.7 mM potassium chloride
  • 1.25 mM calcium chloride
  • 1.20 mM magnesium sulfate
  • 1.20 mM monopotassium phosphate
  • 25 mM sodium bicarbonate
  • 10 mM HEPES
  • 11.1 mM D-glucose