Kjeldahl Copper Catalyst Tablets, Technical, Contains 1g Na2SO4 And Equivalent Of 0.1g CuSO4, Fisher Chemical™
Fisher Chemical™
Kjeldahl Copper Catalyst Tablets, Technical, Contains 1g Na2SO4 And Equivalent Of 0.1g CuSO4, Fisher Chemical™
Kjeldahl Copper Catalyst Tablets, Technical, Contains 1g Na2SO4 And Equivalent Of 0.1g CuSO4, Fisher Chemical, Quantity: 1000 tablets, Blue-White, MDL Number: 147336, Packaging: Plastic powder jar, Physical Form: Solid
Quantity:
1000 Tablets
500 Tablets
Catalog number K-0120-81
also known as K/0120/81
Price (JPY)
-
Quantity:
1000 Tablets
Request bulk or custom format
Specifications
ColorBlue-White
MDL Number147336
PackagingPlastic powder jar
Physical FormSolid
Quantity1000 Tablets
View more
Kjeldahl Copper Catalyst Tablets, Technical, Contains 1g Na2SO4 And Equivalent Of 0.1g CuSO4, Fisher Chemical, Quantity: 1000 tablets, Blue-White, MDL Number: 147336, Packaging: Plastic powder jar, Physical Form: Solid