Search
Search
| Catalog Number | Quantity |
|---|---|
| K534000 also known as K5340-00 | 1 Kit |
The ViraPower™ HiPerform™ Lentiviral FastTiter™ Gateway™ Expression Kit includes all the components needed to generate lentivirus, including vector kit, 293FT cell line, and the support kit. This kit combines Invitrogen's ViraPower™ HiPerform™ Lentiviral and Gateway™ technologies to facilitate easy recombination-based cloning and lentiviral-based, high-level expression of a target gene in dividing and non-dividing mammalian cells. The pLenti7.3/V5-DEST™ vector is equipped with two key genetic elements, making it a HiPerform™ vector: the Woodchuck Posttranscriptional Regulatory Element (WPRE) and the central Polypurine Tract (cPPT) sequence from the HIV-1 integrase gene to produce at least 4-fold increase in protein expression compared to vectors lacking these elements. In addition, the vector allows for an accurate determination of titer of functional lentivirus in just two days using Emerald Green Fluorescent Protein (EmGFP).
Advantages
Key Features
Kit includes
pLenti7.3/V5-DEST™ Gateway™ Vector Kit
• pLenti7.3/V5-DEST™ Gateway™ Vector (150 ng/μL), 40 μL
• pLenti7.3/V5-GW/lacZ Control Vector (0.5 μg/μL) , 20 μlL
Store at -20°C.
ViraPower™ Lentiviral Support Kit
Contains enough reagents for 20 transfections in 10 cm tissue culture dishes:
• Lyophilized Packaging Mix, 195 μg, store at -20°C
• Lipofectamine™ 2000, 0.75 mL, store at 4°C
One Shot™ Stbl3™ chemically competent E. coli box
• One Shot™ Competent Cells, 20 single-use aliquots of 50 μL
• S.O.C. medium
• pUC19 supercoiled plasmid control
Store at -80°C.
293FT cell line
• 3 x 106 frozen cells in 1 mL of Freezing Medium, store in liquid nitrogen.