Lactose Monohydrate, Certified AR for Analysis, Fine Powder, Fisher Chemical™
Lactose Monohydrate, Certified AR for Analysis, Fine Powder, Fisher Chemical™
Lactose Monohydrate, Certified AR for Analysis, Fine Powder, Fisher Chemical™
Lactose Monohydrate, Certified AR for Analysis, Fine Powder, Fisher Chemical™
Fisher Chemical™

Lactose Monohydrate, Certified AR for Analysis, Fine Powder, Fisher Chemical™

Quantity:
500 g
1 kg
Catalog number L-0250-60
also known as L/0250/60
Price (JPY)
-
Contact Us ›
Quantity:
1 kg
Request bulk or custom format
Specifications
CAS64044-51-5
ColorWhite
Formula Weight360.3g/mol
IUPAC Name(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate
InChI KeyWSVLPVUVIUVCRA-RJMJUYIDSA-N
View more
Lactose Monohydrate, Certified AR for Analysis, Fine Powder