Benzo[c]cinnoline, 99%, Thermo Scientific Chemicals
Benzo[c]cinnoline, 99%, Thermo Scientific Chemicals
Benzo[c]cinnoline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzo[c]cinnoline, 99%, Thermo Scientific Chemicals

 
Catalog NumberQuantity
L00409.031 g
Catalog number L00409.03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS230-17-1
IUPAC Namebenzo[c]cinnoline
Molecular FormulaC12H8N2
InChI KeySWJXWSAKHXBQSY-UHFFFAOYSA-N
SMILESC1=CC=C2C(=C1)N=NC1=CC=CC=C21
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)Yellow or grey-green
FormCrystals or crystalline powder
Melting Point (clear melt)153-161?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only