1-Undecyne, 97%
1-Undecyne, 97%
1-Undecyne, 97%
Thermo Scientific Chemicals

1-Undecyne, 97%

CAS: 2243-98-3 | C11H20 | 152.281 g/mol
Quantity:
10 g
50 g
Catalog number L02883.18
also known as L02883-18
Price (JPY)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS2243-98-3
IUPAC Nameundec-1-yne
Molecular FormulaC11H20
InChI KeyYVSFLVNWJIEJRV-UHFFFAOYSA-N
SMILESCCCCCCCCCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0% (UK-sourced material)
Identification (FTIR)Conforms (UK-sourced material)
Refractive Index1.4285-1.4350 @ 20?C (UK-sourced material)
FormLiquid
Assay from Supplier's CofA≥96.0% (US-sourced material)
View more
1-Undecyne is used as a reagent organic synthesis. Further, it is used in the preparation of an antimalarial and antitrypanosomal agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Undecyne is used as a reagent organic synthesis. Further, it is used in the preparation of an antimalarial and antitrypanosomal agent.

Solubility
Miscible with acetone, ether, ethanol and benzene. Slightly miscible with water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Listunov, D.; Billot, C.; Joly, E.; Fabing, I.; Volovenko, Y.; Génisson, Y.; Maraval, V.; Chauvin, R. Extended structural modulation of bio-inspired chiral lipidic alkynylcarbinols as antitumor pharmacophores. Tetrahedron 2015, 71 (41), 7920-7930.
  2. Wu, H.; Kelley, C. J.; Pino-Figueroa, A.; Vu, H. D.; Maher, T. J. Macamides and their synthetic analogs: Evaluation of in vitro FAAH inhibition. Bioorg. Med. Chem. 2013, 21 (17), 5188-5197.