Benzenesulfonyl fluoride, 98%
Benzenesulfonyl fluoride, 98%
Benzenesulfonyl fluoride, 98%
Thermo Scientific Chemicals

Benzenesulfonyl fluoride, 98%

CAS: 368-43-4 | C6H5FO2S | 160.16 g/mol
Quantity:
1 g
5 g
Catalog number L04689.06
also known as L04689-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS368-43-4
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Supplier's CofA≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only