1-Phenylpyrrolidine, 98+%
1-Phenylpyrrolidine, 98+%
1-Phenylpyrrolidine, 98+%
Thermo Scientific Chemicals

1-Phenylpyrrolidine, 98+%

CAS: 4096-21-3 | C10H13N | 147.22 g/mol
Quantity:
5 g
25 g
Catalog number L05239.14
also known as L05239-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS4096-21-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange to brown
Assay (GC)≥98.0%
Refractive Index1.5800-1.5840 @ 20?C
FormLiquid
1-Phenylpyrrolidine are used as flooding intestinal coccidiosis, flooding Xiao NET intermediate.It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenylpyrrolidine are used as flooding intestinal coccidiosis, flooding Xiao NET intermediate.It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents. Store cold 2-8°C.
RUO – Research Use Only