Methyl 3-phenylpropionate, 98%
Methyl 3-phenylpropionate, 98%
Methyl 3-phenylpropionate, 98%
Thermo Scientific Chemicals

Methyl 3-phenylpropionate, 98%

CAS: 103-25-3 | C10H12O2 | 164.204 g/mol
Quantity:
50 g
250 g
Catalog number L05310.18
also known as L05310-18
Price (JPY)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS103-25-3
IUPAC Namemethyl 3-phenylpropanoate
Molecular FormulaC10H12O2
InChI KeyRPUSRLKKXPQSGP-UHFFFAOYSA-N
SMILESCOC(=O)CCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5005-1.5045 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Methyl 3-phenylpropionate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 3-Phenylpropionic Acid Methyl Ester is an ester derivative of 3-phenylpropionic acid that is sometimes present in fragrance ingredients. It is also used as an application, aromatic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-phenylpropionate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 3-Phenylpropionic Acid Methyl Ester is an ester derivative of 3-phenylpropionic acid that is sometimes present in fragrance ingredients. It is also used as an application, aromatic intermediate.

Solubility
Soluble in chloroform. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. X Lu.; Z Xu.; G Yang. Process development of the Sharpless catalytic asymmetric dihydroxylation reaction to prepare methyl (2 R, 3 S)-2, 3-dihydroxy-3-phenylpropionate. Organic Process Research & Development. 2000, 4,(6), 575-576.
  2. K Sasaki.; T Sakakura.; Y Tokunaga.; K Wada. C= C Double bond insertion in catalytic CH activation. Dehydrogenative cross coupling of arenes with olefins. Chemistry Letters. 1988, 17,(4), 685-688.