4-Methyl-1-pentyn-3-ol, 97%
4-Methyl-1-pentyn-3-ol, 97%
4-Methyl-1-pentyn-3-ol, 97%
Thermo Scientific Chemicals

4-Methyl-1-pentyn-3-ol, 97%

CAS: 565-68-4 | C6H10O | 98.145 g/mol
Catalog NumberQuantity
L05318.18
also known as L05318-18
50 g
Catalog number L05318.18
also known as L05318-18
Price (JPY)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS565-68-4
IUPAC Name4-methylpent-1-yn-3-ol
Molecular FormulaC6H10O
InChI KeyUTIFIONYBLSHIL-UHFFFAOYNA-N
SMILESCC(C)C(O)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Refractive Index1.4335-1.4385 @ 20?C
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to yellow
4-Methyl-1-pentyn-3-ol, is used as standard Pharmaceutical intermediate and also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methyl-1-pentyn-3-ol, is used as standard Pharmaceutical intermediate and also used in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. J Boukouvalas.; S Côté.; B Ndzi. Facile access to 4-(1-alkynyl)-2 (5H)-furanones by Sonogashira coupling of terminal acetylenes with β-tetronic acid bromide: efficient synthesis of cleviolide. Tetrahedron Letters. 2007, 48,(1), 105-107.
  2. SF Kirsch.; LE Overman.; NS White. Catalytic Asymmetric Synthesis of Allylic Aryl Ethers. Organic letters. 2007, 48,(1), 105-107.