2-(4-Ethoxyphenyl)ethanol, 98%
2-(4-Ethoxyphenyl)ethanol, 98%
2-(4-Ethoxyphenyl)ethanol, 98%
Thermo Scientific Chemicals

2-(4-Ethoxyphenyl)ethanol, 98%

CAS: 22545-15-9 | C10H14O2 | 166.22 g/mol
Quantity:
2 g
10 g
Catalog number L05597.04
also known as L05597-04
Price (JPY)
-
Quantity:
2 g
Request bulk or custom format
Chemical Identifiers
CAS22545-15-9
IUPAC Name2-(4-ethoxyphenyl)ethan-1-ol
Molecular FormulaC10H14O2
InChI KeyCNMVSNTVPZWQMI-UHFFFAOYSA-N
SMILESCCOC1=CC=C(CCO)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥97.5%
Melting Point (clear melt)38.5-44.5?C
FormCrystals or powder or crystalline powder or flakes
2-(4-Ethoxyphenyl)ethanol, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(4-Ethoxyphenyl)ethanol, is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. A Jarrahpour.; M Zarei. Synthesis of novel N-(4-ethoxyphenyl) azetidin-2-ones and their oxidative N-deprotection by ceric ammonium nitrate. Molecules . 2007, 12,(10), 2364-2379.
  2. YJ Lee.; SD Shukla. Pro-and anti-apoptotic roles of c-Jun N-terminal kinase (JNK) in ethanol and acetaldehyde exposed rat hepatocytes. European journal of pharmacology . 2005, 508,(1-3), 31-45.