4-Fluorobenzamide, 98%
4-Fluorobenzamide, 98%
4-Fluorobenzamide, 98%
4-Fluorobenzamide, 98%
Thermo Scientific Chemicals

4-Fluorobenzamide, 98%

CAS: 824-75-9 | C7H6FNO | 139.129 g/mol
Quantity:
5 g
25 g
100 g
Catalog number L06831.22
also known as L06831-22
Price (JPY)
-
Request A Quote
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS824-75-9
IUPAC Name4-fluorobenzamide
Molecular FormulaC7H6FNO
InChI KeyVNDHYTGVCGVETQ-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or crystalline powder
Melting Point (clear melt)154-161?C
Appearance (Color)White to pale brown
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only