Naphthalene-2-carboxamide, 98%
Naphthalene-2-carboxamide, 98%
Naphthalene-2-carboxamide, 98%
Naphthalene-2-carboxamide, 98%
Thermo Scientific Chemicals

Naphthalene-2-carboxamide, 98%

CAS: 2243-82-5 | C11H9NO | 171.199 g/mol
Quantity:
2 g
10 g
Catalog number L08391.04
also known as L08391-04
Price (JPY)
-
Request A Quote
Quantity:
2 g
Request bulk or custom format
Chemical Identifiers
CAS2243-82-5
IUPAC Namenaphthalene-2-carboxamide
Molecular FormulaC11H9NO
InChI KeyJVXXKQIRGQDWOJ-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder
Assay (HPLC)≥97.5%
Melting Point (clear melt)191.5-197.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only