6,7-Dimethoxyquinazoline-2,4-dione, 98%
6,7-Dimethoxyquinazoline-2,4-dione, 98%
6,7-Dimethoxyquinazoline-2,4-dione, 98%
Thermo Scientific Chemicals

6,7-Dimethoxyquinazoline-2,4-dione, 98%

CAS: 28888-44-0 | C10H10N2O4 | 222.20 g/mol
Quantity:
10 g
50 g
Catalog number L08411.18
also known as L08411-18
Price (JPY)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS28888-44-0
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
FormPowder
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only