Thermo Scientific Chemicals

N-Phenyliminodiacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: L08437.06
5 g, Each
Thermo Scientific Chemicals

N-Phenyliminodiacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: L08437.06
5 g, Each
Quantity
Catalog number: L08437.06
also known as L08437-06
Price (JPY)

Chemical Identifiers

CAS
1137-73-1
IUPAC Name
2-[(carboxymethyl)(phenyl)amino]acetic acid
Molecular Formula
C10H11NO4
InChI Key
GQBWTAGIANQVGB-UHFFFAOYSA-N
SMILES
OC(=O)CN(CC(O)=O)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder or granules
Appearance (Color)
White to cream to yellow to pale green or pale purple to pale pink to gray
Assay (HPLC)
≥96.0%
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text