2,4,5-Trichlorophenyl isothiocyanate, 98%
2,4,5-Trichlorophenyl isothiocyanate, 98%
2,4,5-Trichlorophenyl isothiocyanate, 98%
Thermo Scientific Chemicals

2,4,5-Trichlorophenyl isothiocyanate, 98%

CAS: 23165-46-0 | C7H2Cl3NS | 238.51 g/mol
Quantity:
1 g
5 g
Catalog number L11551.06
also known as L11551-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS23165-46-0
IUPAC Name1,2,4-trichloro-5-isothiocyanatobenzene
Molecular FormulaC7H2Cl3NS
InChI KeyPJLRSYLEFZNICX-UHFFFAOYSA-N
SMILESClC1=CC(Cl)=C(C=C1Cl)N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder
Assay (GC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only