2,6-Dibromo-4-nitrotoluene, 95%
2,6-Dibromo-4-nitrotoluene, 95%
2,6-Dibromo-4-nitrotoluene, 95%
2,6-Dibromo-4-nitrotoluene, 95%
Thermo Scientific Chemicals

2,6-Dibromo-4-nitrotoluene, 95%

CAS: 110127-07-6 | C7H5Br2NO2
Quantity:
5 g
25 g
Catalog number L13102.14
also known as L13102-14
Price (JPY)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS110127-07-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
FormCrystals or powder or crystalline powder or fused/lumpy solid
Melting Point (clear melt)50.0-59.0?C
Assay (GC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only