1,4-Cyclohexanebis(methylamine), cis + trans, 96%
1,4-Cyclohexanebis(methylamine), cis + trans, 96%
1,4-Cyclohexanebis(methylamine), cis + trans, 96%
Thermo Scientific Chemicals

1,4-Cyclohexanebis(methylamine), cis + trans, 96%

CAS: 2549-93-1 | C8H18N2 | 142.246 g/mol
Quantity:
1 g
5 g
25 g
Catalog number L13407.03
also known as L13407-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS2549-93-1
IUPAC Name[4-(azaniumylmethyl)cyclohexyl]methanaminium
Molecular FormulaC8H20N2
InChI KeyOXIKYYJDTWKERT-UHFFFAOYSA-P
SMILES[NH3+]CC1CCC(C[NH3+])CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4890-1.4950 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only