Methyl 3-(3-pyridyl)propionate, 98%
Methyl 3-(3-pyridyl)propionate, 98%
Methyl 3-(3-pyridyl)propionate, 98%
Thermo Scientific Chemicals

Methyl 3-(3-pyridyl)propionate, 98%

CAS: 84199-98-4 | C9H11NO2 | 165.192 g/mol
Quantity:
1 g
5 g
Catalog number L14590.03
also known as L14590-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Beilstein132852
CAS84199-98-4
Chemical Name or MaterialMethyl 3-(3-pyridyl)propionate
EINECS Number000-000-0
Formula Weight165.19
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only