Fumaronitrile, 98+%
Fumaronitrile, 98+%
Fumaronitrile, 98+%
Thermo Scientific Chemicals

Fumaronitrile, 98+%

CAS: 764-42-1 | C4H2N2 | 78.074 g/mol
Quantity:
1 g
5 g
Catalog number L14911.03
also known as L14911-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS764-42-1
IUPAC Name(2E)-but-2-enedinitrile
Molecular FormulaC4H2N2
InChI KeyKYPOHTVBFVELTG-OWOJBTEDSA-N
SMILESN#C\C=C\C#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only